missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF146 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA551908
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: LSQQRIYSGE NPFACKVCGK VFSHKSNLTE HEHFHTREKP FECNECGKAF SQKQYVIKHQ NTHTGEKLFE CNECGKSFSQ KENLLTHQKI HTGEKPFECK DCGKAFIQKS NLIR Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.
ZNF146 (zinc finger protein 146), also known as OZF (only zinc finger protein), is a 292 amino acid nuclear protein that binds DNA and belongs to the Kruppel C2H2-type zinc-finger protein family. ZNF146 interacts with UBC9 (ubiquitin carrier protein 9) and RAP1 (repressor/activator protein 1 homolog), and is highly expressed in multiple pancreatic and colorectal cancers. Lower levels of expression have been observed in kidney, lung, placenta and brain.
Spécification
| ZNF146 | |
| Polyclonal | |
| Unconjugated | |
| ZNF146 | |
| Only zinc finger protein; OZF; Zinc finger protein 146; zinc finger protein OZF; ZNF146 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 102643033, 103691098, 7705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15072 | |
| LOC102643033, ZNF146 | |
| Recombinant protein corresponding to Human ZNF146. Recombinant protein control fragment (Product #RP-100741). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu