missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF546 Polyclonal specifically detects ZNF546 in Human samples. It is validated for Western Blot.
Spécification
Spécification
| Antigène | ZNF546 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formule | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Numéro d’ordre du gène | NP_848639 |
| Alias de gène | MGC43537, zinc finger protein 49, zinc finger protein 546, ZNF49 |
| Symboles de gène(s) | ZNF546 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?