missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25849-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a uridine 5′ -monophosphate synthase. The encoded protein is a bifunctional enzyme that catalyzes the final two steps of the de novo pyrimidine biosynthetic pathway. The first reaction is carried out by the N-terminal enzyme orotate phosphoribosyltransferase which converts orotic acid to orotidine-5′ -monophosphate. The terminal reaction is carried out by the C-terminal enzyme OMP decarboxylase which converts orotidine-5′ -monophosphate to uridine monophosphate. Defects in this gene are the cause of hereditary orotic aciduria. Alternate splicing results in multiple transcript variants.Spécification
ACD | |
Polyclonal | |
Unconjugated | |
ACD | |
ACD, PIP1, PTOP, TINT1, TPP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65057 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96AP0 | |
ACD | |
ACD Fusion Protein Ag23021 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |