missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMDEC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17899-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
ADAMDEC1 may play an important role in the control of the immune response and during pregcy.Spécification
ADAMDEC1 | |
Polyclonal | |
Unconjugated | |
ADAMDEC1 | |
ADAM DEC1, ADAM like protein decysin 1, ADAM like, decysin 1, ADAMDEC1, M12.219 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27299 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15204 | |
ADAMDEC1 | |
ADAMDEC1 Fusion Protein Ag12265 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |