missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTSL5 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Marque: Novus Biologicals NBP2-41120
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ADAMTSL5 Polyclonal specifically detects ADAMTSL5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| ADAMTSL5 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ADAMTSL5 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ADAMTSL5. The immunogen is located within the last 50 amino acids of ADAMTSL5. Amino Acid Squence: YARPWSPAEDSRIRLTA | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgY |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| ADAMTSL-5, ADAMTS-like 5, ADAMTS-like protein 5, thrombospondin type-1 domain-containing protein 6, thrombospondin, type I, domain containing 6, THSD6 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 339366 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu