Learn More
ADORA2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 21071-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17251366
  • 140.00€ / 20µL
Expédition estimée: 24-05-2024
Description
The ADORA2B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ADORA2B. This antibody recognizes human, mouse antigen. The ADORA2B antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
ADORA2B
Polyclonal
Unconjugated
20 μL
136
Liquid
Western Blot, ELISA
600 μg/mL
Rabbit
Primary
Human, Mouse