Learn More
ADORA2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21071-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Description
The ADORA2B antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human ADORA2B. This antibody recognizes human, mouse antigen. The ADORA2B antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
ADORA2B | |
Polyclonal | |
Unconjugated | |
20 μL | |
136 | |
Liquid |
Western Blot, ELISA | |
600 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |