missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRA2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14266-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.Spécification
| ADRA2A | |
| Polyclonal | |
| Unconjugated | |
| ADRA2A | |
| ADRA2, ADRA2A, ADRA2R, ADRAR, Alpha 2A adrenergic receptor, Alpha 2A adrenoceptor, Alpha 2A adrenoreceptor, Alpha 2AAR, ALPHA2AAR, ZNF32 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11551, 150, 25083 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08913, P22909, Q01338 | |
| Adra2a | |
| ADRA2A Fusion Protein Ag5562 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu