missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRA2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Tekniske data
| Antigène | ADRA2A |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Produktkode | Brand | Quantité | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantité | Pris | Antal & tilgængelighed | |||||
|
16812834
|
Proteintech
14266-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
16802834
|
Proteintech
14266-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Beskrivelse
Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.Tekniske data
| ADRA2A | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P08913, P22909, Q01338 | |
| 11551, 150, 25083 | |
| ADRA2A Fusion Protein Ag5562 | |
| Primary | |
| -20°C | |
| ADRA2A |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ADRA2, ADRA2A, ADRA2R, ADRAR, Alpha 2A adrenergic receptor, Alpha 2A adrenoceptor, Alpha 2A adrenoreceptor, Alpha 2AAR, ALPHA2AAR, ZNF32 | |
| Adra2a | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel