Learn More
Clinitrol™ 290 Reference Solution, 2mL; Advanced Instruments
Critical normal human serum midpoint of 290mOsm
Marque: Advanced Instruments 3MA029
Description
Critical normal human serum midpoint of 290mOsm
- Checks performance of freezing-point osmometers in the critical area of normal serum values
- Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
- Pipetted into a tube and run just like normal serum
- Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
- Does not require mixing, reconstituting or refrigerating; one-year shelf life
Suggestions de produits
Customers who viewed this item also viewed.
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.