missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17299-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Descripción
Aspartylglucosaminidase is involved in the catabolism of N-linked oligosaccharides of glycoproteins. It cleaves asparagine from N-acetylglucosamines as one of the final steps in the lysosomal breakdown of glycoproteins. The lysosomal storage disease aspartylglycosaminuria is caused by a deficiency in the AGA enzyme. Alternatively spliced transcript variants have been identified.Especificaciones
AGA | |
Polyclonal | |
Unconjugated | |
AGA | |
AGA, AGU, aspartylglucosaminidase, ASRG, GA, Glycosylasparaginase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11593, 175, 290923 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P20933, P30919, Q64191 | |
Aga | |
AGA Fusion Protein Ag10681 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |