missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13907-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The type II cAMP-dependent protein kinase (PKA) is a multifunctional kinase with a broad range of substrates. Specificity of PKA signaling is mediated by the compartmentalization of the kinase to specific sites within the cell. To maintain this specific localization, the R subunit (RII) of PKA interacts with specific RII-anchoring proteins, designated A-kinase anchoring proteins (AKAP). AKAP 3, also known as AKAP 110, FSP95, PRKA3 and SOB1, binds both PKA and PDE4A and functions as a scaffolding protein in spermatozoa to regulate local cAMP concentrations and modulate sperm functions. Expression of AKAP 3 in normal tissues is restricted to the testis, where bicarbonate stimulates tyrosine phosphorylation of AKAP 3, thereby increasing its recruitment of PKA. AKAP-3 also exhibits high expression in patients with epithelial ovarian cancer (EOC). It demonstrates tumor-restricted expression and appears to be associated with worse overall survival, which make AKAP 3 a potential target for antigen-specific immunotherapy in EOC.Spécification
AKAP3 | |
Polyclonal | |
Unconjugated | |
AKAP3 | |
A kinase anchor protein 3, AKAP 110, AKAP 3, AKAP110, AKAP3, Cancer/testis antigen 82, CT82, Fibrousheathin 1, Fibrousheathin I, FSP95, PRKA3, SOB1, Sperm oocyte binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10566, 11642, 312720 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75969, O88987 | |
Akap3 | |
AKAP3 Fusion Protein Ag4885 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |