missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALMS1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27231-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
THRAP3 is a 955 amino acid novel nuclear receptor and a member of the Thyroid hormone receptor associated proteins with a LISH domain and no obvious known sequence motifs. THRAP3 is a 150 kDa subunit of the large multiprotein complex TRAP and is important in the maintece of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It is also known to play an important role in transcriptional coactivation. It is known to interact with DKC1, nop5/nap65 and fibrillarin. From the Northern blot analysis of several human tissues it was found that TRAP150 is ubiquitously expressed.Spécification
ALMS1 | |
Polyclonal | |
Unconjugated | |
ALMS1 | |
ALMS1, ALSS, Alstrom syndrome 1, Alstrom syndrome protein 1, KIAA0328 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7840 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TCU4 | |
ALMS1 | |
ALMS1 Fusion Protein Ag26100 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |