Learn More
Angiopoietin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23302-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart.Spécification
Angiopoietin 1 | |
Polyclonal | |
Unconjugated | |
ANGPT1 | |
AGP1, AGPT, ANG 1, ANG1, angiopoietin 1, ANGPT1, KIAA0003 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11600, 284, 89807 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08538, O35460, Q15389 | |
ANGPT1 | |
Angiopoietin 1 Fusion Protein Ag18829 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |