missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25153-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The specific function of this gene has not yet been determined, however, it is associated with the plasma membrane of undifferentiated, proliferating endothelial cells and differentiated villus enterocytes. Alternatively spliced transcript variants encoding different isoforms have been identified.Spécification
Annexin A13 | |
Polyclonal | |
Unconjugated | |
ANXA13 | |
Annexin 13, annexin A13, Annexin XIII, ANX13, ANXA13, Intestine specific annexin, ISA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
312, 69787 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P27216, Q99JG3 | |
ANXA13 | |
Annexin A13 Fusion Protein Ag18581 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |