missing translation for 'onlineSavingsMsg'
Learn More

Annexin A2 Mouse anti-Human, Mouse, Clone: 3H1B11, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 60051-1-IG-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837005

  • 121.38€ / 20µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

Annexin A2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P07355, P07356
Anxa2
Annexin A2 Fusion Protein Ag1777
20 μL
Primary
Human, Mouse
Antibody
IgG2b
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin)
3H1B11
Unconjugated
ANXA2
Annexin 2, annexin A2, Annexin II, ANX2, ANX2L4, ANXA2, CAL1H, Calpactin 1 heavy chain, Calpactin I heavy chain, Chromobindin 8, LIP2, Lipocortin II, LPC2, LPC2D, P36, PAP IV, Protein I
Mouse
Antigen Affinity Chromatography
RUO
12306, 302
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions