missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A3 Mouse anti-Human, Clone: 1A11E4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66405-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.Spécification
Annexin A3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12429 | |
ANXA3 | |
Annexin A3 Fusion Protein Ag2381 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin) | |
1A11E4 | |
Unconjugated | |
ANXA3 | |
35 alpha calcimedin, Annexin 3, annexin A3, Annexin III, ANX3, ANXA3, Lipocortin III, PAP III | |
Mouse | |
Protein A | |
RUO | |
306 | |
-20°C | |
Liquid |