missing translation for 'onlineSavingsMsg'
Learn More

ANP32B Mouse anti-Human, Clone: 5A1F12, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66160-1-IG-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837785

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity.
Spécification

Spécification

ANP32B
Monoclonal
0.89 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q92688
ANP32B
ANP32B Fusion Protein Ag1274
20 μL
Primary
Human
Antibody
IgG2b
Immunofluorescence, Immunocytochemistry, Western Blot
5A1F12
Unconjugated
ANP32B
ANP32B, APRIL, PHAPI2, Silver stainable protein SSP29, SSP29
Mouse
Protein A
RUO
10541
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions