Learn More
Caveolin 1 Mouse, Unlabeled, Clone: C060, BD
Mouse Monoclonal Antibody
394.00€ - 620.00€
Spécification
Antigène | Caveolin 1 |
---|---|
Clone | C060 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15875229
|
BD Biosciences
610057 |
50 μg |
394.00€
1 kit |
Expédition estimée: 07-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15885229
|
BD Biosciences
610058 |
150 μg |
620.00€
1 kit |
Expédition estimée: 07-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Identified as a tyrosine phosphorylated protein in Rous sarcoma virus-transformed chick embryo fibroblasts (CEF), caveolin is now known to be ubiquitously expressed. Caveolin (also known as VIP21) localizes to non-clathrin membrane invaginations (caveolae) on the inner surface of the plasma membrane. This transmembrane protein plays a structural role in these specializations. Caveolin is also present at the trans-Golgi network (TGN) and similar quantities are found in apically and basolaterally destined transport vesicles. Caveolin is part of a complex containing glycosylphosphatidylinositol (GPI)-linked molecules and cytoplasmic signaling proteins. Caveolin is a transmembrane adaptor molecule that can simultaneously recognize GPI-linked proteins and interact with downstream cytoplasmic signaling molecules, such as c-yes, Annexin II, and hetero-trimeric G proteins. Caveolin-1 can generate two forms, α and β, due to alternate splicing of the mRNA. Caveolin-1 forms large lipid-binding homo-oligomers which are believed to play a role in caveolae formation. It may also function as a scaffolding protein which concentrates and organizes signaling molecules, a role supported by the fact that caveolin-1 interacts directly with inactive Ras and G-protein α subunits.
Host Species: Mouse
Clone: C060
Isotype: IgM
Species Reactivity: Human
Immunogen: Human Caveolin 1 aa. 1-97
Formula Weight [Chemical]: 22kDa
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spécification
Caveolin 1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
C060 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rabbit, Rat | |
IgM | |
Affinity Purified |
For Research Use Only.