Learn More
CD15 Mouse anti-Human, FITC, Clone: HI98 (also known as HIM1), BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 555401
Informations supplémentaires : Poids : 0.02000kg
Description
Reacts with 3-fucosyl-N-acetyllactosamine (3-FAL), a 220kDa carbohydrate structure, also called X-hapten, SSEA-1, CD15 or Lewis X. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and to a varying degree on monocytes, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. This antibody is also suitable for staining formalin-fixed, paraffin-embedded tissue sections without pretreatment. Since the Abs are recognizing a carbohydrate epitope (3-fucosyl-N-acetyllactosamine) they also should work across species and not only for human.
Flow Cytometry
Spécification
CD15 | |
Monoclonal | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HI98 (also known as HIM1) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Mononuclear Cells | |
100 Tests | |
Immunology | |
Human | |
IgM κ |
For Research Use Only.