Learn More
IGF1R alpha Monoclonal Antibody (1H7), PE, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA516465
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
IGF1R alpha Monoclonal antibody specifically detects IGF1R alpha in Human samples. It is validated for Flow Cytometry.Spécification
IGF1R alpha | |
Monoclonal | |
Conc. Not Determined | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P08069 | |
Igf1r | |
purified human placental IGF-I receptor | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
1H7 | |
PE | |
Igf1r | |
A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Mouse | |
Protein G | |
RUO | |
3480 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |