Learn More
CD326 Mouse anti-Human, APC, Clone: EBA-1, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 347200
Informations supplémentaires : Poids : 0.03000kg
Description
Anti-EpCAM, clone EBA-1, is derived from the fusion of SP2 myeloma cells with spleen cells from BALB/c mice immunized with macromolecular aggregates containing breast carcinoma–associated mucin BCA-225.
Anti-EpCAM is directed against a human epithelial adhesion molecule (EpCAM) that has been identified as a ∽40-kilodalton (kd) protein in Western blotting. This antigen is also known as epithelial surface antigen (ESA) and epithelial glycoprotein 2 (EGP-2).
Spécification
CD326 | |
Monoclonal | |
12μg/mL | |
EpCAM | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 λ |
Flow Cytometry | |
EBA-1 | |
APC | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Breast carcinoma-associated mucin BCA-225 | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
For Research Use Only.