Learn More
CD47 Rat anti-Mouse, FITC, Clone: MIAP301, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 561890
Informations supplémentaires : Poids : 0.01000kg
Description
The miap301 monoclonal antibody specifically binds to the extracellular domain of CD47, also known as Integrin-Associated Protein (IAP). CD47 is a 50-kDa plasma membrane protein with an extracellular immunoglobulin variable region-like domain, a multiple membrane-spanning segment, and a short C-terminal cytoplasmic tail. IAP is often physically associated with β3 integrins, and it may act as a transducer element in activation mediated by these integrins. CD47 is expressed by a wide variety of cell types, including some cells lacking integrins, such as erythrocytes. The cytoplasmic tail region is expressed as alternatively spliced forms, which are differentially expressed in diverse tissues, suggesting that IAP may have distinct functions in various tissues. CD47 and its ligand, CD172a, are proposed to mediate bi-directional signaling to modify neural synaptic activity and regulate the phagocytic activities of macrophages.
Spécification
CD47 | |
Monoclonal | |
0.5mg/mL | |
Cd47; IAP; Integrin-associated protein; Itgp | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
MIAP301 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not reported | |
0.1 mg | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.