Learn More
CD80 Mouse anti-Human, BB515, Clone: L307.4, BD
Mouse Monoclonal Antibody
184.00€ - 386.00€
Spécification
Antigène | CD80 |
---|---|
Clone | L307.4 |
Applications | Flow Cytometry, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Conjugué | Brilliant Blue 515 (BB515) |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15874019
|
BD Biosciences
565009 |
25 Tests |
184.00€
1 pièce |
Expédition estimée: 06-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15864019
|
BD Biosciences
565008 |
100 Tests |
386.00€
1 pièce |
Expédition estimée: 06-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The L307.4 monoclonal antibody specifically binds to B7/BB1, a 60 kDa transmembrane glycoprotein that was clustered as CD80 in the Fifth International Workshop on Human Leukocyte Differentiation Antigens. CD80, a member of the Ig supergene family, is expressed on activated B cells, T cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells, CD28 and CD152 (CTLA-4). CD80 is also expressed on activated CD4-positive and CD8-positive T cells, appearing late after activation suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 by anti-CD28 or by CD80 results in T-cell activation and a signal for IL-2 production.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Spécification
CD80 | |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Brilliant Blue 515 (BB515) | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
IgG1 κ | |
Affinity Purified | |
B7.1; B7-1; Activation B7-1 antigen; B7; BB1; CD28LG; CD28LG1; LAB7 |
L307.4 | |
Monoclonal | |
Mouse | |
Human | |
Human B7-transfected L cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.