missing translation for 'onlineSavingsMsg'
Learn More

Cyclin B Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 610219

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 15845339

  • ONLINE EXCLUSIVE PRICE
    219.00€
    218.28€ / 1 kit
    Économisez 0.72€
    0.33% de réduction

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

The B-type cyclins serve as subunits of the major mitotic protein kinase in many different organisms. In mammals, Cyclin B is a subunit for the cdc2 (Cdk1) protein kinase and is synthesized during late S and G2 phases. Cyclin B associates with inactive cdc2 and facilitates phosphorylation of cdc2 at Thr-14 and Tyr-15. This phosphorylation maintains the inactive state until the end of G2. The inactive Cyclin B-cdc2 complex continues to accumulate in the cytoplasm until the CDC25 phosphatase dephosphorylates Thr-14 and Tyr-15, rendering the complex active at the G2/M boundary. This mitotic kinase complex remains active until the metaphase/anaphase transition when Cyclin B becomes degraded. This degradation process appears to be ubiquitin-dependent and is necessary for the cell to exit mitosis. Also, the conserved domain in B-type cyclins, known as the P box, is required for CDC25 activation.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Spécification

Spécification

Cyclin B
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Human, Rat
IgG1
Western Blot
18
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.