Learn More
HAX-1 Mouse, Unlabeled, Clone: 52, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 610825
Informations supplémentaires : Poids : 0.01000kg
Description
Cell-cell interactions in the immune system, mediated by membrane-bound antigen receptors, trigger a cascade of signaling mechanisms collectively known as the T-cell response. Upon antigen binding, cellular tyrosine kinases are activated resulting in the phosphorylation of HS1 or Lck-binding protein 1 (LckBP1), which is involved in thymic negative selection. HS1 is one of the earliest intracellular proteins to be tyrosine phosphorylated in response to antigen-receptor cross-linking in T cells. HAX-1 (HS1-associated protein X-1) is a 35kDa protein found associated with HS1 by co-immunoprecipitation assays and the two-hybrid system. HAX-1 is ubiquitously expressed and localized in the mitochondria, endoplasmic reticulum, and the nuclear envelope. The HS1-HAX-1 complex may be critical for signal transduction pathways initiated by antigen presentation to T cells.
Immunofluorescence, Immunoprecipitation, Western Blotting
Spécification
HAX-1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Human HAX-1 aa. 10-148 | |
150 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
52 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.