Learn More
HLA-DR Mouse anti-Human, BUV661, Clone: G46-6, BD
Mouse Monoclonal Antibody
292.74€ - 591.60€
Spécification
Antigène | HLA-DR |
---|---|
Clone | G46-6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | Brilliant Ultraviolet 661 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15884059
|
BD Biosciences
565074 |
25 Tests |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
15874059
|
BD Biosciences
565073 |
100 Tests |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
The G46-6 monoclonal antibody specifically binds to HLA-DR, a major histocompatibility complex (MHC) class II antigen. HLA-DR antigens are encoded by genes within the Human Leukocyte Antigen (HLA) Complex located on chromosome 6. HLA-DR is a transmembrane heterodimeric glycoprotein composed of an α chain (36 kDa) and a β subunit (27 kDa) expressed primarily on antigen presenting cells: B cells, dendritic cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in mediating cellular interactions during antigen presentation to CD4-positive T cells.
For optimal and reproducible results, BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant dyes are used in the same experiment. Fluorescent dye interactions may cause staining artifacts which may affect data interpretation. The BD Horizon Brilliant Stain Buffer was designed to minimize these interactions. More information can be found in the Technical Data Sheet of the BD Horizon Brilliant Stain Buffer (Cat. No. 563794).
Due to spectral differences between labeled cells and beads, using BD™ CompBeads can result in incorrect spillover values when used with BD Horizon BUV661 reagents. Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended. Different BUV661 reagents (eg, CD4 vs. CD45) can have slightly different fluorescence spillover therefore, it may also be necessary to use clone-specific compensation controls when using these reagents.
Spécification
HLA-DR | |
Flow Cytometry | |
Brilliant Ultraviolet 661 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
G46-6 | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG2a κ | |
Affinity Purified | |
MHC class II antigen; HLA class II histocompatibility antigen |
For Research Use Only.