missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Mouse anti-Human, Brilliant Violet 605, Clone: G46-6, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 562845

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15872289

  • 710.00€ / 1 pièce
Expédition estimée: 17-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The G46-6 monoclonal antibody specifically binds to HLA-DR, a major histocompatibility complex (MHC) class II antigen. HLA-DR antigens are encoded by genes within the Human Leukocyte Antigen (HLA) Complex located on chromosome 6. HLA-DR is a transmembrane heterodimeric glycoprotein composed of an α chain (36 kDa) and a β subunit (27 kDa) expressed primarily on antigen presenting cells: B cells, dendritic cells, monocytes, macrophages, and thymic epithelial cells. HLA-DR is also expressed on activated T cells. This molecule plays a major role in mediating cellular interactions during antigen presentation to CD4-positive T cells.

For optimal and reproducible results, BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant dyes are used in the same experiment. Fluorescent dye interactions may cause staining artifacts which may affect data interpretation. The BD Horizon Brilliant Stain Buffer was designed to minimize these interactions. More information can be found in the Technical Data Sheet of the BD Horizon Brilliant Stain Buffer (Cat. No. 563794).

Spécification

Spécification

HLA-DR
Monoclonal
Brilliant Violet 605
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Flow Cytometry
G46-6
MHC class II antigen; HLA class II histocompatibility antigen
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.