Learn More
Anti-Human CD81 (5A6), Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 65195-1-IG-500UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD81 antibody from Proteintech is a mouse monoclonal antibody generated with human oci-ly8 cell line of human CD81. This antibody recognizes human antigen. The CD81 antibody has been validated for the following applications: FC analysis.Spécification
| CD81 | |
| Monoclonal | |
| 0.5 mg/mL | |
| S5.7; TAPA1; TSPAN28 | |
| Human OCI-LY8 cell line | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 5A6 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 975 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |