Learn More
IL-17A Mouse anti-Human, Alexa Fluor 488, Clone: N49-653, BD
Mouse Monoclonal Antibody
265.00€ - 590.00€
Spécification
Antigène | IL-17A |
---|---|
Clone | N49-653 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | Alexa Fluor 488 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15880239
|
BD Biosciences
560489 |
25 Tests |
265.00€
1 pièce |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15870239
|
BD Biosciences
560488 |
100 Tests |
590.00€
1 pièce |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Human IL-17A, also known as IL-17, is a proinflammatory cytokine that is encoded by the IL17A gene in chromosome 6. IL-17A is produced as a disulfide-linked homodimer comprised of two mature 136-amino acid polypeptides. It is a member of the IL-17 family of structurally related cytokines, designated IL-17A through IL-17F. Activated memory T cells, especially Th17 cells (specialized IL-17A-producing CD4+ T cells distinct from Th1 and Th2 cells) produce IL-17 and provide protective immunity against pathogens. Activated CD8+ T cells, γδT cells, NK cells and neutrophils can also be activated to produce IL-17A. IL-17A binds to and exerts its biological activity through IL-17 receptors (IL-17R) that are expressed by a variety of target cells including fibroblasts, epithelial and endothelial cells, monocytes/macrophages and mast cells. The ubiquitous IL-17R expression pattern may explain the broad tissue responsiveness to IL-17. IL-17 induces stromal cells to secrete cytokines and chemokines involved in inflammatory and hematopoietic processes. For example, IL-17 induces fibroblasts to produce IL-6, IL-8, G-CSF and express increased surface ICAM-1. The N49-653 antibody reacts with human IL-17A.
Host Species: Mouse
Clone: N49-653
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IL-17A Recombinant Protein
Intracellular Staining
Spécification
IL-17A | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Human | |
Human IL-17A Recombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
N49-653 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
IL-17; CTLA8; Cytotoxic T-lymphocyte-associated serine esterase 8 |
For Research Use Only.