missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3 Monoclonal Antibody (2F2), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA120048
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
MA1-20048 detects IL-3 in human samples.
IL-3 Monoclonal antibody specifically detects IL-3 in Human samples. It is validated for ELISA.Spécification
IL-3 | |
Monoclonal | |
3.6 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P08700 | |
IL3 | |
Recombinant, full length, human IL-3 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA | |
2F2 | |
Unconjugated | |
IL3 | |
BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF | |
Mouse | |
Protein G | |
RUO | |
3562 | |
4° C | |
Liquid |