missing translation for 'onlineSavingsMsg'
Learn More

IL-4 Rat anti-Mouse, Unlabeled, Clone: 11B11, BD

Rat Monoclonal Antibody

Marque:  BD Biosciences 554434

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15847698

  • 499.00€ / 1 pièce
Expédition estimée: 23-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The 11B11 antibody reacts with mouse interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 from PMA-stimulated EL-4 supernatant. This is a neutralizing antibody.

Host Species: Rat
Clone: 11B11
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Partially Purified Mouse IL-4

ELISA

Spécification

Spécification

IL-4
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Partially Purified Mouse IL-4
0.5 mg
Immunology
Mouse
IgG1
ELISA
11B11
Unconjugated
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.