Learn More
IMPDH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA527792
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (95%), Rat (82%), Xenopus laevis (88%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IMPDH1 Polyclonal antibody specifically detects IMPDH1 in Human samples. It is validated for Western Blot.Spécification
IMPDH1 | |
Polyclonal | |
Unconjugated | |
IMPDH1 | |
B930086D20Rik; DKFZp781N0678; HAMAP-Rule:MF_03156}; IMP (inosine 5'-monophosphate) dehydrogenase 1; IMP (inosine monophosphate) dehydrogenase 1; IMP dehydrogenase 1; IMP dehydrogenase 1 {ECO:0000255; IMPD; IMPD 1; IMPD 1 {ECO:0000255; IMPD1; IMPDH 1; IMPDH 1 {ECO:0000255; Impdh1; IMPDH-I; inosine 5'-phosphate dehydrogenase 1; inosine monophosphate dehydrogenase 1; inosine-5'-monophosphate dehydrogenase 1; inosine-5'-monophosphate dehydrogenase 1 {ECO:0000255; LCA11; RP10; sWSS2608 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P20839 | |
IMPDH1 | |
Recombinant fragment corresponding to a region within amino acids 61 and 310 of Human IMPDH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |