Learn More
Anti-Mouse CD69 (H1.2F3), Proteintech
Hamster Monoclonal Antibody
Marque: Proteintech 65105-1-IG-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CD69 antibody from Proteintech is a armenian hamster monoclonal antibody generated with mouse dendritic epidermal t cell line y245 of mouse CD69. This antibody recognizes mouse antigen. The CD69 antibody has been validated for the following applications: FC analysis.Spécification
| CD69 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Armenian Hamster | |
| Affinity purification | |
| RUO | |
| 12515 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| H1.2F3 | |
| Unconjugated | |
| Mouse dendritic epidermal T cell line Y245 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Flow Antibody | |
| IgG |