missing translation for 'onlineSavingsMsg'
Learn More

serine hydroxymethyltransferase 1 (soluble) (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant SHMT1.

Marque:  Abnova H00006470-A01.50uL

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00010kg

Code produit. 16175235

  • 226.00€ / 50µL
Expédition estimée: 08-07-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes the cellular form of serine hydroxymethyltransferase, a pyridoxal phosphate-containing enzyme that catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon units for synthesis of methionine, thymidylate, and purines in the cytoplasm. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing of this gene results in 2 transcript variants encoding 2 different isoforms. Additional transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Sequence: EKVLEACSIACNKNTCPGDRSALRPSGLRLGTPALTSRGLLEKDFQKVAHFIHRGIELTLQIQSDTGVRATLKEFKERLAGDKYQAAVQALREEVESFASLFPLPGLPD
Spécification

Spécification

serine hydroxymethyltransferase 1 (soluble)
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant SHMT1.
SHMT1
CSHMT/MGC15229/MGC24556/SHMT
Mouse
50 μL
Yes
6470
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_004169
SHMT1
SHMT1 (NP_004160, 374 a.a. ∼ 482 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions