missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARCL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA543150
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
Peptide sequence: QESITKREEN QEQPRNYSHH QLNRSSKHSQ GLRDQGNQEQ DPNISNGEEE Sequence homology: Human: 100%
SPARCL1 Polyclonal antibody specifically detects SPARCL1 in Human samples. It is validated for Western Blot.Spécification
SPARCL1 | |
Polyclonal | |
Unconjugated | |
SPARCL1 | |
Ecm2; Extracellular matrix protein 2; hevin; High endothelial venule protein; MAST 9; MAST9; matrix glycoprotein Sc1; PIG33; proliferation-inducing protein 33; SC1; SPARC like 1; SPARCL1; SPARC-like 1; SPARC-like 1 (hevin); SPARC-like 1 (mast9, hevin); SPARC-like protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8404 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14515 | |
SPARCL1 | |
Synthetic peptide directed towards the N-terminal of human SPARCL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |