missing translation for 'onlineSavingsMsg'
Learn More

serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin), Mouse, Clone: 4G3, Abnova™

Mouse monoclonal antibody raised against a partial recombinant SPINLW1.

Marque:  Abnova H00057119-M02A.200uL

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00010kg

Code produit. 16179006

  • 326.00€ / 200µL
Expédition estimée: 08-07-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes an epididymal protease inhibitor, which contains both kunitz-type and WAP-type four-disulfide core (WFDC) protease inhibitor consensus sequences. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene is a member of the WFDC gene family and belongs to the telomeric cluster. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq

Sequence: PGLTDWLFPRRCPKIREECEFQERDVCTKDRQCQDNKKCCVFSCGKKCLDLKQDVCEMPKETGPCLAYFLHWWYDKKDNTCSMFVYGGCQGNNNNFQSKANCLNTCKNKRFP
Spécification

Spécification

serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin)
Monoclonal
Unconjugated
ascites with no preservative
NM_020398
SPINLW1
SPINLW1 (NP_065131, 22 a.a. ∼ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
Primary
Human
Antibody
IgM κ
ELISA, Western Blot
4G3
Mouse monoclonal antibody raised against a partial recombinant SPINLW1.
SPINLW1
EPPIN/EPPIN1/EPPIN2/EPPIN3/WAP7/WFDC7/dJ461P17.2
Mouse
200 μL
Yes
57119
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Ascites
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions