Learn More
AQP7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 25131-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16833675
  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
Description
Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.
Spécification
Spécification
AQP7
Polyclonal
Unconjugated
AQP7
AQP 7, AQP7, AQP7L, AQP9, AQPap, Aquaglyceroporin 7, aquaporin 7, Aquaporin 7 like, Aquaporin adipose
Rabbit
Antigen Affinity Chromatography
RUO
11832, 364
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14520, O54794
AQP7
AQP7 Fusion Protein Ag17945
150 μL
Primary
Mouse, Human