Learn More
Aquaporin-4 Antibody (AQP4/3323), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-44612-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Spécification
| Aquaporin-4 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| AQP4/3323 | |
| Unconjugated | |
| aquaporin 4, MGC22454 | |
| Human recombinant Aquaporin-4 protein fragment (around aa200-323) (Uniprot: P55087). Exact sequence is proprietary. | |
| 100 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 361 | |
| Store at 4⪚C. | |
| IgG1 κ |