missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL8A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17060-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
May play a role in lysosomes motility. Alternatively, may play a role in chromosome segregation.Spécification
ARL8A | |
Polyclonal | |
Unconjugated | |
ARL8A | |
ARL10B, ARL8A, GIE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
127829, 68724 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VEH3, Q96BM9 | |
Arl8a | |
ARL8A Fusion Protein Ag10788 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |