Learn More
ARPC1B Mouse anti-Human, Unconjugated, Clone: 2C8A1, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67320-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829885
  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 
Description
Description
The ARPC1B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARPC1B. This antibody recognizes human, rat antigen. The ARPC1B antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
ARPC1B
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat
Western Blot, ELISA
2C8A1
Unconjugated
20 μL
10095
IgG1