missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arrestin C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | Arrestin C |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16847953
|
Proteintech
11100-2-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16837953
|
Proteintech
11100-2-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Arrestin-C, also known as retinal cone Arrestin-3, X-Arrestin or cArr, is a member of the Arrestin family of proteins. It is predomitly found in the retina and pineal gland and localizes to the inner and outer segments of red-, green- and blue-cone photoreceptors and the inner plexiform regions. Two Arrestin-C isoforms exist due to alternative splicing. Isoform 1 is the mature full length protein and isoform 2 is truncated, ending with an arginine for amino acid residue 359. Arrestin-C expression is stimulated by retinoic acid. It may play a role in retina-specific signal transduction and bind to photoactivated-phosphorylated red/green opsins. In addition, Arrestin-C forms homodimers and oligomers with beta-Arrestins and may regulate beta-Arrestin mediated signaling.Spécification
Arrestin C | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P36575, Q9EQP6 | |
170735, 407 | |
Arrestin C Fusion Protein Ag1580 | |
Primary | |
-20°C | |
ARR3 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARR3, Arrestin C, ARRX, C arrestin, CAR, cArr, Cone arrestin, Retinal cone arrestin 3, X arrestin | |
ARR3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |