missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASF1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22258-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody specifically reacts with the 19 kDa ASF1B protein.
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The encoded protein is the substrate of the tousled-like kinase family of cell cycle-regulated kinases, and may play a key role in modulating the nucleosome structure of chromatin by ensuring a constant supply of histones at sites of nucleosome assembly.Spécification
ASF1B | |
Polyclonal | |
Unconjugated | |
ASF1B | |
ASF1B, CCG1 interacting factor A II, CIA II, hAsf1, hAsf1b, hCIA II, Histone chaperone ASF1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55723 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NVP2 | |
ASF1B | |
ASF1B Fusion Protein Ag17763 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |