missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASK1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28201-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mitogen-activated protein (MAP) kinase cascades are activated in response to various extracellular stimuli, including cytokines, growth factors and environmental stresses. A novel MAP kinase kinase kinase (MAPKKK) was recently identified and designated ASK1 (for apoptosis signal-regulating kinase 1) and MAPKKK5. ASK1 activated two different subgroups of MAPKK, MKK4 and MKK6, which in turn activated c-Jun N-terminal kinase (JNK) and p38 MAP kinase, respectively. ASK1/MAPKKK5 is activated by TNFR and Fas through the interaction with members of the TRAF family and Fas-associated protein Daxx. Overexpression of ASK1 induced apoptotic cell death, and a catalytically inactive form of ASK1 inhibited TNF-a-induced apoptosis. ASK1 is expressed in variety of human and mouse tissues.Spécification
ASK1 | |
Polyclonal | |
Unconjugated | |
MAP3K5 | |
ASK 1, ASK1, MAP3K5, MAPK/ERK kinase kinase 5, MAPKKK5, MEK kinase 5, MEKK 5, MEKK5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26408, 4217 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35099, Q99683 | |
Map3k5 | |
ASK1 Fusion Protein Ag28092 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |