missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP12A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13231-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Catalyzes the hydrolysis of ATP coupled with the exchange of H(+) and K(+) ions across the plasma membrane. Responsible for potassium absorption in various tissues.Spécification
ATP12A | |
Polyclonal | |
Unconjugated | |
ATP12A | |
ATP12A, ATP1AL1, Proton pump | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
479 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P54707 | |
ATP12A | |
ATP12A Fusion Protein Ag4105 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |