missing translation for 'onlineSavingsMsg'
Learn More

ATP1A3 (C-terminal) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10868-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827543

  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is generated against the C-terminal region (665-1013aa) of ATP1A3 and detects the band around 100-113 kDa in SDS-PAGE.

The sodium/potassium ATPase is an integral membrane enzyme found in all cells of higher organisms and is responsible for the ATP-dependent transport of sodium and potassium across the cell membrane. This membrane-bound enzyme is related to a number of other ATPases including sarcoplasmic and endoplasmic reticulum calcium ATPase (SERCA) and plasma membrane calcium ATPase (PMCA). The sodium/potassium ATPase consists of a large, multipass, transmembrane catalytic subunit, termed the alpha subunit, and an associated smaller glycoprotein, termed the beta subunit. Studies indicate that there are three isoforms of the alpha subunit (alpha 1, alpha 2, alpha 3) and two isoforms of the beta subunit (beta 1 and beta 2) encoded by two multigene families. The different isoforms of the sodium/potassium ATPase exhibit tissue-specific and developmental patterns of expression. The alpha 1 and beta mRNAs are present in all cell types examined, whereas the alpha 2 and alpha 3 mRNAs exhibit a more restricted pattern of cell-specific expression. The alpha-3 subunit has been found in neuronal and to skeletal and cardiac muscle, lung and stomach tissues.
Spécification

Spécification

ATP1A3 (C-terminal)
Polyclonal
Unconjugated
ATP1A3
ATP1A3, DYT12, RDP, Sodium pump subunit alpha 3
Rabbit
Antigen Affinity Chromatography
RUO
232975, 24213, 478
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P06687, P13637, Q6PIC6
ATP1A3
ATP1A3 (C-terminal) Fusion Protein Ag1313
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions