missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUTS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25001-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
AUTS2 also named as FBRSL2 or autism susceptibility candidate 2 is a 1259 amino acid protein, which belongs to the AUTS2 family. AUTS2 is highly expressed in brain, kidney and skeletal muscle. AUTS2 exists as four isoforms produced by alternative splicing events and is encoded by a gene located on human chromosome 7. The molecular weight of AUTS2 isoform is 65-70 kDa in the brain. The functional role of AUTS2 is not well known, although some studies have identified a putative role in transcriptional regulation during neuronal development.Spécification
AUTS2 | |
Polyclonal | |
Unconjugated | |
AUTS2 | |
AUTS2, FBRSL2, KIAA0442 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26053, 319974, 498173 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A0A087WPF7, Q8WXX7 | |
Auts2 | |
AUTS2 Fusion Protein Ag21853 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |