Learn More
Axl Mouse anti-Human, Super Bright 436, Clone: DS7HAXL, eBioscience™
Mouse Monoclonal Antibody
Marque: Invitrogen 62-1087-42
Informations supplémentaires : Poids : 0.01000kg
Description
This DS7HAXL monoclonal antibody recognizes human Axl, a member of the TAM family of tyrosine kinase receptors that also includes MerTK and Tyro3. This DS7HAXL antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. It has also been shown to stain formalin-fixed, paraffin-embedded tissues for IHC analysis. Applications Reported: This DS7HAXL antibody has been reported for use in flow cytometric analysis. Applications Tested: This DS7HAXL antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser
Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. Axl is a member of the TAM receptor family, which also includes also MerTK and TYRO3. Axl contains protein kinase activity and is activated by dimerization and auto-phosphorylation upon ligand binding. In response to its ligands, Protein S and GAS6, Axl activates anti-inflammatory pathways (through SOCS-1 and SOCS-3), which limits TLR and cytokine signaling. This results in dampened inflammatory responses in macrophages and DCs. TAM receptors are also involved in phagocytosis of apoptotic cells. Mice lacking all three TAM receptors have several degenerative phenotypes linked to inefficient removal of apoptotic cells and membranes (e.g., in the retina and the male reproductive tract) and develop a severe autoimmune phenotype akin to systemic lupus erythematosus, including the production of broad spectrum auto-antibodies. In addition, Axl may function as a putative entry receptor for filoviruses. Axl is also used to identify a specific subpopulation of human blood dendritic cells, also referred to as Siglec 6+/Axl+ dendritic cells. Cellular expression of Axl can be upregulated by TLR ligands, such as LPS or poly I:C. Soluble Axl is generated by proteolytic cleavage of the membrane form. Increased plasma levels may be indicative of inflammation and cancer.Spécification
Axl | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P30530 | |
AXL | |
Affinity chromatography | |
RUO | |
558 | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
DS7HAXL | |
Super Bright 436 | |
AXL | |
ARK; AXL; JTK11; Tyro7; UFO | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Liquid |