Learn More
B-Raf Antibody (CL4004) - Azide and BSA Free, Novus Biologicals™
Code produit. 30300434 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantité unitSize
30300434 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit. 30300434 Fournisseur Novus Biologicals Code fournisseur NBP344118
Mouse Monoclonal Antibody
B-Raf Monoclonal antibody specifically detects B-Raf in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY
Spécification
Antigène B-Raf
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone CL4004
Concentration LYOPH
Conjugué Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formule Lyophilized in 5% Trehalose.
Alias de gène 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109
Espèces hôtes Mouse
Immunogène This antibody was generated using a recombinant protein sequence of P15056, with the exact immunogen sequence remaining proprietary.
Méthode de purification Protein A purified
Quantité 100 μL
État réglementaire RUO
Disciplines de recherche Apoptosis, Phospho Specific
Primaire ou secondaire Primary
Identification génétique (Entrez) 673
Reconstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Espèces cibles Human
Contenu et stockage Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG2b