Learn More
Novus Biologicals™ B7-H3/CD276 Antibody (enoblituzumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28383-50ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| B7-H3/CD276 | |
| Monoclonal | |
| Unconjugated | |
| Q5ZPR3 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| enoblituzumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule | |
| B7-H3/ CD276 | |
| 50 μg | |
| Immunology | |
| 80381 | |
| Human | |
| Purified |