Learn More
B7-H3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14453-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16833104
  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
Description
CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.
Spécification
Spécification
B7-H3
Polyclonal
Unconjugated
CD276
4Ig B7 H3, B7 homolog 3, B7-H3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule, UNQ309/PRO352
Rabbit
Antigen Affinity Chromatography
RUO
80381
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5ZPR3
CD276
B7-H3 Fusion Protein Ag6006